Learn More
Medchemexpress LLC Fluorene-d8 | 35782-21-9 | 174.27 | C13H2D8 | 100 MG
SDP
Catalog No. 5000301460
Encompass_Preferred
Fluorene-d8 is the deuterium-labeled isotopologue of fluorene supplied as an analytical reference standard for laboratory research. It is intended for use in mass spectrometry, tracer studies, and synthetic applications where a deuterium-labeled fluorene is required.
  • Deuterium-labeled reference for mass spectrometry and tracer studies.
  • Helps improve quantitation accuracy as an internal standard.
  • Suitable for analytical method development and validation.
  • Useful in synthetic chemistry as a labeled building block.
  • Stable molecular formula C13H2D8 and molecular weight 174.27 g/mol.
  • Provided in small packaged quantities for laboratory use.
Catalog No. 50-003-01460 Supplier Medchemexpress LLC Supplier No. HYW026772S1100MG
Fluorene-d8 is the deuterium-labeled isotopologue of fluorene supplied as an analytical reference standard for laboratory research. It is intended for use in mass spectrometry, tracer studies, and synthetic applications where a deuterium-labeled fluorene is required.
  • Deuterium-labeled reference for mass spectrometry and tracer studies.
  • Helps improve quantitation accuracy as an internal standard.
  • Suitable for analytical method development and validation.
  • Useful in synthetic chemistry as a labeled building block.
  • Stable molecular formula C13H2D8 and molecular weight 174.27 g/mol.
  • Provided in small packaged quantities for laboratory use.