Learn More
Medchemexpress LLC Fluorene-d8 | 35782-21-9 | 174.27 g·mol⁻¹ | C13H2D8 | 1 G
SDP
Catalog No. 5000301236
Encompass_Preferred
Fluorene-d8 is the deuterium-labeled analogue of fluorene used as an isotopically labeled standard for analytical, tracer, and mechanistic studies. It is typically supplied as a solid with molecular formula C13H2D8 and molecular weight 174.27 g·mol⁻¹. Intended uses include mass spectrometry internal standards, metabolite tracing, and isotopic labeling experiments; consult the Certificate of Analysis for lot-specific purity and storage conditions.
  • Deuterium-labeled internal standard for MS analyses.
  • Useful for isotopic tracing and metabolic studies.
  • Molecular formula C13H2D8 and molecular weight 174.27 g·mol⁻¹.
  • Supplied in multiple pack sizes including 1 g.
  • Certificate of Analysis provides lot-specific purity and storage instructions.
Catalog No. 50-003-01236 Supplier Medchemexpress LLC Supplier No. HYW026772S11G
Fluorene-d8 is the deuterium-labeled analogue of fluorene used as an isotopically labeled standard for analytical, tracer, and mechanistic studies. It is typically supplied as a solid with molecular formula C13H2D8 and molecular weight 174.27 g·mol⁻¹. Intended uses include mass spectrometry internal standards, metabolite tracing, and isotopic labeling experiments; consult the Certificate of Analysis for lot-specific purity and storage conditions.
  • Deuterium-labeled internal standard for MS analyses.
  • Useful for isotopic tracing and metabolic studies.
  • Molecular formula C13H2D8 and molecular weight 174.27 g·mol⁻¹.
  • Supplied in multiple pack sizes including 1 g.
  • Certificate of Analysis provides lot-specific purity and storage instructions.