Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC Fluorenone | 486-25-9 | 100.0% | 180.20 | 1 KG
SDP

Catalog No. 5000400489
Encompass_Preferred

9H-fluoren-9-one is a biochemical reagent that can be used as a biological material or organic compound for life science related research. It is a solid with a light yellow to yellow appearance.

  • Formula: C13H8O
  • Molecular weight: 180.20
  • Purity: 99.97%
  • Shipping: Room temperature in continental US
  • Storage: Store at room temperature for 3 years; in solvent at -80°C for 2 years or -20°C for 1 year
  • Solubility (in vitro): DMSO: 100 mg/mL (554.94 mM)

Catalog No. 50-004-00489 Supplier Medchemexpress LLC Supplier No. HY321811KG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

9H-fluoren-9-one is a biochemical reagent that can be used as a biological material or organic compound for life science related research. It is a solid with a light yellow to yellow appearance.

  • Formula: C13H8O
  • Molecular weight: 180.20
  • Purity: 99.97%
  • Shipping: Room temperature in continental US
  • Storage: Store at room temperature for 3 years; in solvent at -80°C for 2 years or -20°C for 1 year
  • Solubility (in vitro): DMSO: 100 mg/mL (554.94 mM)

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.