Learn More
Medchemexpress LLC Fluorenone-d8 | 137219-34-2 | 188.25 | 5 MG
SDP
Catalog No. 5000401118
Encompass_Preferred
Fluorenone-d8 is a deuterium-labeled compound, with eight hydrogen atoms in the fluorenone structure replaced by deuterium. It has the molecular formula C13D8O and a molecular weight of 188.25. This compound is intended for research use only, primarily as a laboratory chemical and for the manufacture of substances.
  • Deuterium-labeled compound
  • Solid at room temperature
  • Melting point 93-95°C
  • Identified for laboratory chemical use
  • Suitable for substance manufacturing
  • Stable as powder for 3 years at -20°C
  • Stable as powder for 2 years at 4°C
  • Stable in solvent for 6 months at -80°C
  • Stable in solvent for 1 month at -20°C
Catalog No. 50-004-01118 Supplier Medchemexpress LLC Supplier No. HY32181S5MG
Fluorenone-d8 is a deuterium-labeled compound, with eight hydrogen atoms in the fluorenone structure replaced by deuterium. It has the molecular formula C13D8O and a molecular weight of 188.25. This compound is intended for research use only, primarily as a laboratory chemical and for the manufacture of substances.
  • Deuterium-labeled compound
  • Solid at room temperature
  • Melting point 93-95°C
  • Identified for laboratory chemical use
  • Suitable for substance manufacturing
  • Stable as powder for 3 years at -20°C
  • Stable as powder for 2 years at 4°C
  • Stable in solvent for 6 months at -80°C
  • Stable in solvent for 1 month at -20°C