Learn More
Medchemexpress LLC Fluorenone-d8 | 137219-34-2 | 98% | 188.25 | 1 MG
SDP
Catalog No. 5000401905
Encompass_Preferred
Fluorenone-d8 is a labeled analogue of 9-Fluorenone, a polycyclic aromatic ketone. It is primarily identified as a laboratory chemical for substance manufacturing and is intended for research use only, not validated for medical applications. This product should be handled by experienced and qualified scientists in appropriately equipped facilities.
  • Possesses photosensitizing properties
  • Can undergo electrochemical polymerization
  • Utilized as an initiator in visible-light cured resin systems
  • For research use only
Catalog No. 50-004-01905 Supplier Medchemexpress LLC Supplier No. HY32181S1MG
Fluorenone-d8 is a labeled analogue of 9-Fluorenone, a polycyclic aromatic ketone. It is primarily identified as a laboratory chemical for substance manufacturing and is intended for research use only, not validated for medical applications. This product should be handled by experienced and qualified scientists in appropriately equipped facilities.
  • Possesses photosensitizing properties
  • Can undergo electrochemical polymerization
  • Utilized as an initiator in visible-light cured resin systems
  • For research use only