Learn More
Abcam Fluorescein mono-beta-D-N-acetylglucosamine, 5MG
Catalog No. 5051309
Encompass_Preferred
MW 535.5 Da, Purity >95%. Fluorogenic substrate for hexosaminidase enzymes. Used for enzymatic characterization of O-GlcNAcase isoforms producing the fluorophore fluorescein upon enzyme activity, with a green emission spectrum (~520 nm).
The product is subject to the following: Abcam Restricted Use Statement
Catalog No. 50-513-09 Supplier Abcam Supplier No. AB2748785MG
MW 535.5 Da, Purity >95%. Fluorogenic substrate for hexosaminidase enzymes. Used for enzymatic characterization of O-GlcNAcase isoforms producing the fluorophore fluorescein upon enzyme activity, with a green emission spectrum (~520 nm).
The product is subject to the following: Abcam Restricted Use Statement