Learn More
Abcam Fluorescein mono-ß-D-galactopyranoside, 5MG
Supplier:  Abcam AB2750885MG
Encompass_Preferred
MW 494.4 Da, Purity >90%. Useful for enzymatic analyses of β-galactosidase where 1st order kinetics are desired.
The product is subject to the following: Abcam Restricted Use Statement
Catalog No. 50-996-067