Learn More
Cayman Chemical Fluorescamine, Purity: ≥95%, CAS: 38183-12-9, 50 mg
SDP
Catalog No. 502947023
Encompass_Preferred
An amine-reactive fluorescent probe; has commonly been used as a reagent in fluorometric protein assays; has commonly been used as a reagent in fluorometric protein assays; displays excitation/emission maxima of 370/488 nm, respectively, in acetone
Catalog No. 50-294-7023 Supplier Cayman Chemical Supplier No. 3428650 MG
An amine-reactive fluorescent probe; has commonly been used as a reagent in fluorometric protein assays; has commonly been used as a reagent in fluorometric protein assays; displays excitation/emission maxima of 370/488 nm, respectively, in acetone