Learn More
Cayman Chemical FluoresceIn-12-dCTP 25nmol
SDP
Catalog No. 502911831
Encompass_Preferred
A fluorescently labeled form of dCTP; ex/em = 498/517 nm, respectively
Catalog No. 50-291-1831 Supplier Cayman Chemical Supplier No. 3623425 NMOL
A fluorescently labeled form of dCTP; ex/em = 498/517 nm, respectively