Learn More
Cayman Chemical Fluorescein-12-dGTP, Purity: ≥95%, 25 nmol
SDP
Catalog No. 502912038
Encompass_Preferred
A fluorescently labeled form of dGTP; ex/em = 498/517 nm, respectively
Catalog No. 50-291-2038 Supplier Cayman Chemical Supplier No. 3623525 NMOL
A fluorescently labeled form of dGTP; ex/em = 498/517 nm, respectively