Learn More
Cayman Chemical Fluorescein-12-dUTP, Purity: ≥95%, 25 nmol
SDP
Catalog No. 502911830
Encompass_Preferred
A fluorescently tagged form of dUTP; can be used to generate fluorescently labeled DNA via reverse transcription, nick translation, random primed labeling, or PCR; has been used to detect nucleic acids in situ; ex/em = 498/517 nm, respectively
Catalog No. 50-291-1830 Supplier Cayman Chemical Supplier No. 3623225 NMOL
A fluorescently tagged form of dUTP; can be used to generate fluorescently labeled DNA via reverse transcription, nick translation, random primed labeling, or PCR; has been used to detect nucleic acids in situ; ex/em = 498/517 nm, respectively