Learn More
Apexbio Technology LLC Fluorescein-12-dUTP, 25ul (1mM). Cas: 214154-36-6 MFCD: MFCD22200851. Incorporated into DNA/cDNA by PCR, Nick Translation or other application for dye-labeled probe producing.
SDP
Catalog No. 501905497
Encompass_Preferred
Fluorescein-12-dUTP could be used in directly enzymatic labeling of DNA/cDNA, such as PCR, random primed DNA labeling reaction, primer extension, Nick Translation and others. It can be incorporated as a substitute for its natural counterpart dTTP. Other sizes are available. Please inqury us for quote.
Catalog No. 50-190-5497 Supplier Apexbio Technology LLC Supplier No. B82070.025
Fluorescein-12-dUTP could be used in directly enzymatic labeling of DNA/cDNA, such as PCR, random primed DNA labeling reaction, primer extension, Nick Translation and others. It can be incorporated as a substitute for its natural counterpart dTTP. Other sizes are available. Please inqury us for quote.