Learn More
Medchemexpress LLC HY-D0251 1g Medchemexpress, Fluorescein CAS:2321-07-5 Purity:>98%
SDP
Catalog No. 501933163
Encompass_Preferred
Medchemexpress, HY-D0251 1g Fluorescein CAS:2321-07-5 Fluorescein is a synthetic fluorescent tracer. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-193-3163 Supplier Medchemexpress LLC Supplier No. HYD02511G
Medchemexpress, HY-D0251 1g Fluorescein CAS:2321-07-5 Fluorescein is a synthetic fluorescent tracer. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.