Learn More
Cayman Chemical Fluorescein 5(6)-isothiocyanate, Purity: ≥90% (mixture of isomers), CAS: 27072-45-3, 250 mg
SDP
Catalog No. 502947342
Encompass_Preferred
An amine-reactive fluorescent probe; reacts with amine groups to form conjugates with proteins for detection by various fluorescent-based applications; ex/em = 485/535 nm, respectively
Catalog No. 50-294-7342 Supplier Cayman Chemical Supplier No. 33264250 MG
An amine-reactive fluorescent probe; reacts with amine groups to form conjugates with proteins for detection by various fluorescent-based applications; ex/em = 485/535 nm, respectively