Learn More
Cayman Chemical FluoresceIn 5 6-IsothIocyna 1g
SDP
Catalog No. 502948650
Encompass_Preferred
An amine-reactive fluorescent probe; reacts with amine groups to form conjugates with proteins for detection by various fluorescent-based applications; ex/em = 485/535 nm, respectively
Catalog No. 50-294-8650 Supplier Cayman Chemical Supplier No. 332641 G
An amine-reactive fluorescent probe; reacts with amine groups to form conjugates with proteins for detection by various fluorescent-based applications; ex/em = 485/535 nm, respectively