Learn More
Fluorescein-5-EX N-Hydroxysuccinimide Ester, Sigma-Aldrich
Catalog No. 01258843 Shop All MilliporeSigma Supelco Products
Change view
Click to view available options
Quantity:
10 mg
Catalog No. Quantity
01-258-843 10 mg
1 options
Catalog No. 01-258-843 Supplier MilliporeSigma Supelco Supplier No. F955110MG
Add to Cart
Add to Cart
Fluorescein-5-EX N-hydroxysuccinimide ester has been used for the labeling of antibodies.
TRUSTED_SUSTAINABILITY
Specifications
Product Type Fluorescence Reagent
Quantity 10 mg
Chemical Reactivity Carboxylic Acid
Solubility DMF (Dimethylformamide), DMSO (Dimethylsulfoxide)
Labeling Target Antibodies