Learn More
Selleck Chemical LLC Fluorescein-5-isothiocyanate
SDP
Catalog No. 5000002444
Encompass_Preferred
Fluorescein-5-isothiocyanate (FITC Fluorescein isothiocyanate isomer I 5-FITC) is a fluorescent probe capable of being conjugated to tissue and proteins
Catalog No. 50-000-02444 Supplier Selleck Chemical LLC Supplier No. S6928100MG
Fluorescein-5-isothiocyanate (FITC Fluorescein isothiocyanate isomer I 5-FITC) is a fluorescent probe capable of being conjugated to tissue and proteins