Learn More
Cayman Chemical Fluorescein 5-isothiocyanate
SDP
Catalog No. 502937461
Encompass_Preferred
An amine- and thiol-reactive fluorescent probe; forms conjugates with proteins, lipids, and other molecules for detection by a variety of fluorescent-based applications; ex/em = 494/519 nm, respectively
Catalog No. 50-293-7461 Supplier Cayman Chemical Supplier No. 27814250 MG
An amine- and thiol-reactive fluorescent probe; forms conjugates with proteins, lipids, and other molecules for detection by a variety of fluorescent-based applications; ex/em = 494/519 nm, respectively