Learn More
Cayman Chemical Fluorescein 5-isothiocyanate, Purity: ≥90%, CAS: 3326-32-7, 500 mg
SDP
Catalog No. 502941741
Encompass_Preferred
An amine- and thiol-reactive fluorescent probe; forms conjugates with proteins, lipids, and other molecules for detection by a variety of fluorescent-based applications; ex/em = 494/519 nm, respectively
Catalog No. 50-294-1741 Supplier Cayman Chemical Supplier No. 27814500 MG
An amine- and thiol-reactive fluorescent probe; forms conjugates with proteins, lipids, and other molecules for detection by a variety of fluorescent-based applications; ex/em = 494/519 nm, respectively