Learn More
Selleck Chemical LLC Fluorescein-5-isothiocyanate
SDP
Supplier:  Selleck Chemical LLC S692825MG
Encompass_Preferred
Fluorescein-5-isothiocyanate (FITC Fluorescein isothiocyanate isomer I 5-FITC) is a fluorescent probe capable of being conjugated to tissue and proteins
Catalog No. 50-000-02157