Learn More
Cayman Chemical FluoresceIn 5-IsothIocynate 1g
SDP
Supplier:  Cayman Chemical 278141 G
Encompass_Preferred
An amine- and thiol-reactive fluorescent probe; forms conjugates with proteins, lipids, and other molecules for detection by a variety of fluorescent-based applications; ex/em = 494/519 nm, respectively
Catalog No. 50-294-0189