Learn More
Cayman Chemical Fluorescein 5-isothiocyanate, Purity: ≥90%, CAS: 3326-32-7, 5 g
SDP
Catalog No. 502940199
Encompass_Preferred
An amine- and thiol-reactive fluorescent probe; forms conjugates with proteins, lipids, and other molecules for detection by a variety of fluorescent-based applications; ex/em = 494/519 nm, respectively
Catalog No. 50-294-0199 Supplier Cayman Chemical Supplier No. 278145 G
An amine- and thiol-reactive fluorescent probe; forms conjugates with proteins, lipids, and other molecules for detection by a variety of fluorescent-based applications; ex/em = 494/519 nm, respectively