Learn More
Cayman Chemical FluoresceIn-5-maleImIde 10mg
SDP
Catalog No. 502969944
Encompass_Preferred
An activated fluorescent molecule (excitation: 494 nm, emission: 519 nm) used for labeling proteins; reacts optimally with sulfhydryl groups on cysteine side chains at pH 7, forming a stable thioether bond
Catalog No. 50-296-9944 Supplier Cayman Chemical Supplier No. 1638310 MG
An activated fluorescent molecule (excitation: 494 nm, emission: 519 nm) used for labeling proteins; reacts optimally with sulfhydryl groups on cysteine side chains at pH 7, forming a stable thioether bond