Learn More
Aat Bioquest Fluorescein-5-maleimide *CAS 75350-46-8*
SDP
Catalog No. 501952738
Encompass_Preferred
Maleimides are among the most frequently used reagents for thiol modification. In most proteins, the site of reaction is at cysteine residues that either are intrinsically present or resulted from reduction of cystines.
Catalog No. 50-195-2738 Supplier Aat Bioquest Supplier No. 130
Maleimides are among the most frequently used reagents for thiol modification. In most proteins, the site of reaction is at cysteine residues that either are intrinsically present or resulted from reduction of cystines.