Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Fluorescein-5-maleimide 97.0+%, TCI America™
SDP

Catalog No. f081025mg Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
25 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
F081025MG 25 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. F081025MG Supplier TCI America Supplier No. F081025MG
Only null left
Add to Cart
Add to Cart

Chemical Identifiers

CAS 75350-46-8
Molecular Formula C24H13NO7
Molecular Weight (g/mol) 427.368
MDL Number MFCD00077345
InChI Key AYDAHOIUHVUJHQ-UHFFFAOYSA-N
Synonym 5-Maleimidofluorescein
PubChem CID 122038
IUPAC Name 1-(3',6'-dihydroxy-3-oxospiro[2-benzofuran-1,9'-xanthene]-5-yl)pyrrole-2,5-dione
SMILES C1=CC2=C(C=C1N3C(=O)C=CC3=O)C(=O)OC24C5=C(C=C(C=C5)O)OC6=C4C=CC(=C6)O

Specifications

Quantity 25 mg
Formula Weight 427.37
Percent Purity ≥97.0% (HPLC)
Physical Form Crystalline Powder
Chemical Name or Material Fluorescein-5-maleimide
TSCA No
Recommended Storage Refrigerator
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.