Learn More
AdipoGen Fluorescein-5-thiosemicarbazide, AdipoGen Life Sciences. Chemical. CAS: 76863-28-0
SDP
Catalog No. 50596517
Encompass_Preferred
Fluorescein-5-thiosemicarbazide, AdipoGen Life Sciences. Chemical. CAS: 76863-28-0. Formula: C21H15N3O5S. MW: 421.43. Synthetic. Labeling of cell-surface functional groups (glycophorins). Detection of protein carbonyls in aging tissues.
Catalog No. 50-596-517 Supplier AdipoGen Supplier No. CDXF0005M050
Fluorescein-5-thiosemicarbazide, AdipoGen Life Sciences. Chemical. CAS: 76863-28-0. Formula: C21H15N3O5S. MW: 421.43. Synthetic. Labeling of cell-surface functional groups (glycophorins). Detection of protein carbonyls in aging tissues.