Learn More
AdipoGen Fluorescein-5-thiosemicarbazid
SDP
Supplier:  AdipoGen CDXF0005M500
Encompass_Preferred
Chemical. CAS 76863-28-0. Formula C21H15N3O5S. MW 421.43. Synthetic. Labeling of cell-surface functional groups glycophorins. Detection of protein carbonyls in aging tissues.
Catalog No. 50-256-0947