Learn More
AnaSpec FLUORESCEIN-?-AMYLOID (1-42)
Supplier:  AnaSpec AS2352601
Encompass
Beta-Amyloid (1-42), FAM-labeled, Human[FAM-DAEFRHDSGYEVHHQKLVFFAEDVGSNKGAIIGLMVGGVVIA] ; MW 4873.4; (0.1 mg) This is a fluorescent (FAM)-labeled B-Amyloid peptide, Abs/Em=494/521 nm.
Catalog No. 50-824-541