Learn More
Vector Laboratories Avidin DCS Cell Sorting Grade Fluorescein 1 mg
SDP
Catalog No. NC9265088
Encompass
Fluorescein Avidin DCS is a cell sorter grade of Fluorescein Avidin D. Fluorescein Avidin DCS has an optimal F/P ratio and a high fluorescence efficiency. The product is purified to reduce non-specific binding which can hamper resolution in cell sorting and other applications. Although this product has been designed for cell sorting, it can also be used in fluorescent microscopy when higher sensitivity and lower background staining are necessary. This derivative is preferred when using an amplification procedure with Biotinylated Anti-Avidin D. Fluorescein Avidin DCS has an excitation at 495 nm and an emission at about 515 nm.
Catalog No. NC9265088 Supplier Vector Laboratories Supplier No. A20111
Fluorescein Avidin DCS is a cell sorter grade of Fluorescein Avidin D. Fluorescein Avidin DCS has an optimal F/P ratio and a high fluorescence efficiency. The product is purified to reduce non-specific binding which can hamper resolution in cell sorting and other applications. Although this product has been designed for cell sorting, it can also be used in fluorescent microscopy when higher sensitivity and lower background staining are necessary. This derivative is preferred when using an amplification procedure with Biotinylated Anti-Avidin D. Fluorescein Avidin DCS has an excitation at 495 nm and an emission at about 515 nm.