Learn More
R Peptide LLC Fluorescein-Beta-Amyloid (1-42), 0.5 mg
SDP
Supplier:  R Peptide LLC A11191
Encompass
Synthetic peptide, >95%, Store in -20°C. Fluorescein labeled Beta-amyloid peptide (Abeta). Abeta is the major constituent of amyloid plaques in the brains of Alzheimer's patients, and is thought to be the cause of Alzheimer's Disease (AD). AD is the most common neurodegenerative disease and afflicts about 10% of the population over 60
Catalog No. NC0985815