Learn More
Biotium Fluorescein Biotin
Catalog No. 501964784
Encompass_Preferred
Fluorescein biotin (5-((N-(5-(N-(6-(Biotinoyl)amino)hexanoyl)amino)pentyl)thioureidyl) fluorescein) can be used to detect and quantify biotin binding sites and the degree of biotinylation of proteins. Also see biotin-rhodamine 110, biotin-4-fluorescein, and CF Dye Biotin conjugates. Properties: Ex/ Em= 494/518 nm; Yellow solid soluble in DMSO, or DMF; Store at -20°C and protect from light; C42H50N6O8S2; MW: 831; [134759-22-1]
Catalog No. 50-196-4784 Supplier Biotium Supplier No. 80019
Fluorescein biotin (5-((N-(5-(N-(6-(Biotinoyl)amino)hexanoyl)amino)pentyl)thioureidyl) fluorescein) can be used to detect and quantify biotin binding sites and the degree of biotinylation of proteins. Also see biotin-rhodamine 110, biotin-4-fluorescein, and CF Dye Biotin conjugates. Properties: Ex/ Em= 494/518 nm; Yellow solid soluble in DMSO, or DMF; Store at -20°C and protect from light; C42H50N6O8S2; MW: 831; [134759-22-1]