Learn More
Cayman Chemical FluoresceIn BIotIn 5mg
SDP
Catalog No. 503015662
Encompass_Preferred
A fluorescent biotin derivative; an avidin and streptavidin ligand that contains a FITC fluorophore; decreases in fluorescence intensity upon avidin or streptavidin binding have been used to quantify biotin binding stoichiometry
Catalog No. 50-301-5662 Supplier Cayman Chemical Supplier No. 255745 MG
A fluorescent biotin derivative; an avidin and streptavidin ligand that contains a FITC fluorophore; decreases in fluorescence intensity upon avidin or streptavidin binding have been used to quantify biotin binding stoichiometry