Learn More
Biotium Fluorescein CADAVERINE
Catalog No. NC1480931
Encompass_Preferred
Fluorescein cadaverine can be coupled to activated carboxy groups or sulfonyl chlorides.Features: Ex/Em nm (pH 9.0) = 493/517 nm; Yellow solid soluble in DMSO, DMF, or pH>6.5 buffers; Store at 4°C and protect from lightC30H27F6N3O9S; MW: 719.61
Catalog No. NC1480931 Supplier Biotium Supplier No. 92000
Fluorescein cadaverine can be coupled to activated carboxy groups or sulfonyl chlorides.Features: Ex/Em nm (pH 9.0) = 493/517 nm; Yellow solid soluble in DMSO, DMF, or pH>6.5 buffers; Store at 4°C and protect from lightC30H27F6N3O9S; MW: 719.61