Learn More
Medchemexpress LLC Fluorescein di(β-D-galactopyranoside) | 17817-20-8 | MFCD00077344 | ≥98.0% | 656.59 g/mol | C32H32O15 | 10 MG
SDP
Catalog No. 5000245571
Encompass_Preferred
Fluorescein di(β-D-galactopyranoside) is a fluorogenic substrate for β-galactosidase that releases fluorescein upon enzymatic hydrolysis, producing a fluorescent signal suitable for detecting enzyme activity in cells and biochemical assays. Store at -20°C and protect from light; the compound is unstable in solution and should be freshly prepared.
  • Fluorogenic substrate for β-galactosidase.
  • Generates fluorescein with excitation ~488 nm and emission ~520-535 nm.
  • Compatible with flow cytometry, fluorescence microscopy, and plate-reader assays.
  • Molecular formula C32H32O15; molecular weight 656.59 g/mol.
  • Pack sizes suitable for small-scale research use; handle protected from light.
Catalog No. 50-002-45571 Supplier Medchemexpress LLC Supplier No. HY10189510MG
Fluorescein di(β-D-galactopyranoside) is a fluorogenic substrate for β-galactosidase that releases fluorescein upon enzymatic hydrolysis, producing a fluorescent signal suitable for detecting enzyme activity in cells and biochemical assays. Store at -20°C and protect from light; the compound is unstable in solution and should be freshly prepared.
  • Fluorogenic substrate for β-galactosidase.
  • Generates fluorescein with excitation ~488 nm and emission ~520-535 nm.
  • Compatible with flow cytometry, fluorescence microscopy, and plate-reader assays.
  • Molecular formula C32H32O15; molecular weight 656.59 g/mol.
  • Pack sizes suitable for small-scale research use; handle protected from light.