Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC Fluorescein di(β-D-galactopyranoside) | 17817-20-8 | MFCD00077344 | 99.8% | 656.59 | C32H32O15 | 5 MG
SDP

Catalog No. 5000248928
Encompass_Preferred

Fluorescein di(β-D-galactopyranoside) is a fluorogenic substrate that releases fluorescein upon β-galactosidase cleavage, producing a green fluorescent signal for enzymatic assays. It is used for measuring β-galactosidase activity in cell-based and biochemical applications, including senescence-associated β-galactosidase assays.

  • Provides fluorogenic detection of β-galactosidase activity.
  • Produces green fluorescence (Ex 488 nm, Em 520 nm; for SA-βG use Ex 485 nm, Em 535 nm).
  • Soluble in DMSO at 5 mg/mL with ultrasonic warming.
  • Compatible with common in vivo vehicles (PEG300, Tween-80, SBE-B-CD) at reported concentrations.
  • Store at -20 °C and protect from light; unstable in solution-prepare freshly.

Catalog No. 50-002-48928 Supplier Medchemexpress LLC Supplier No. HY1018955MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Fluorescein di(β-D-galactopyranoside) is a fluorogenic substrate that releases fluorescein upon β-galactosidase cleavage, producing a green fluorescent signal for enzymatic assays. It is used for measuring β-galactosidase activity in cell-based and biochemical applications, including senescence-associated β-galactosidase assays.

  • Provides fluorogenic detection of β-galactosidase activity.
  • Produces green fluorescence (Ex 488 nm, Em 520 nm; for SA-βG use Ex 485 nm, Em 535 nm).
  • Soluble in DMSO at 5 mg/mL with ultrasonic warming.
  • Compatible with common in vivo vehicles (PEG300, Tween-80, SBE-B-CD) at reported concentrations.
  • Store at -20 °C and protect from light; unstable in solution-prepare freshly.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.