Learn More
Medchemexpress LLC Fluorescein Di-β-D-Glucuronide | 134869-04-8 | HY-D1460 | 96.6% | 684.55 | 5 MG
SDP
Catalog No. 5000431730
Encompass_Preferred
Fluorescein Di-β-D-Glucuronide is a fluorescent probe used for non-invasive imaging of intestinal bacterial β-glucuronidase activity in nude mice. It reveals the in vitro and in vivo activity of intestinal bacterial β-glucuronidase, facilitating pharmacodynamic studies of specific bacterial β-glucuronidase inhibitors in animal studies.
  • Fluorescent probe
  • Utilized for non-invasive imaging of intestinal bacterial β-glucuronidase activity
  • Reveals in vitro and in vivo activity of intestinal bacterial β-glucuronidase
  • Facilitates pharmacodynamic studies of specific bacterial β-glucuronidase inhibitors
  • Classified as Fluorescent Dye, Enzyme Labeling, Protease Substrate
  • Relevant to Disease Areas: Infection, β-glucuronidase, Bacterial
Catalog No. 50-004-31730 Supplier Medchemexpress LLC Supplier No. HYD14605MG
Fluorescein Di-β-D-Glucuronide is a fluorescent probe used for non-invasive imaging of intestinal bacterial β-glucuronidase activity in nude mice. It reveals the in vitro and in vivo activity of intestinal bacterial β-glucuronidase, facilitating pharmacodynamic studies of specific bacterial β-glucuronidase inhibitors in animal studies.
  • Fluorescent probe
  • Utilized for non-invasive imaging of intestinal bacterial β-glucuronidase activity
  • Reveals in vitro and in vivo activity of intestinal bacterial β-glucuronidase
  • Facilitates pharmacodynamic studies of specific bacterial β-glucuronidase inhibitors
  • Classified as Fluorescent Dye, Enzyme Labeling, Protease Substrate
  • Relevant to Disease Areas: Infection, β-glucuronidase, Bacterial