Learn More
AdipoGen Fluorescein diacetate (10 g)
SDP
Supplier:  AdipoGen CDXF0027G010
Encompass_Preferred
Fluorescein diacetate (10 g), AdipoGen Life Sciences. Chemical. CAS: 596-09-8. Formula: C24H16O7. MW: 416.38. Synthetic. FDA is a non-fluorescent hydrophobic fluorescein derivative that can pass through the cell membrane whereupon intracellular esterases hydrolyze the diacetate group producing the highly fluorescent product fluorescein. The fluorescein molecules accumulate in cells that possess intact membranes so the green fluorescence can be used as a marker of cell viability. Cells that do not possess an intact cell membrane or an active metabolism may not accumulate the fluorescent product and therefore do not exhibit green fluorescence. FDA may be used in combination with PI staining as the non-viable cells take up the PI and stain dead cells red whereas viable cells do not take up the PI and should only stain green.FDA is used in studies on intracellular interactions and membrane permeability, for flow cytometry and as esterase substrate. Spectral data: λex=494nm, λem=521nm.
Catalog No. 50-149-0383