Learn More
Cayman Chemical Fluorescein diacetate, Purity: ≥98%, CAS: 596-09-8, 10 g
SDP
Catalog No. 502910812
Encompass_Preferred
A cell-permeable fluorogenic cell viability probe; cleaved by intracellular esterases to produce fluorescein which displays excitation/emission maxima of 490/520 nm, respectively
Catalog No. 50-291-0812 Supplier Cayman Chemical Supplier No. 3570110 G
A cell-permeable fluorogenic cell viability probe; cleaved by intracellular esterases to produce fluorescein which displays excitation/emission maxima of 490/520 nm, respectively