Learn More
AdipoGen Fluorescein diacetate 5-?maleimide, AdipoGen Life Sciences. Chemical. CAS: 150322-01-3
SDP
Supplier:  AdipoGen CDXF0018M025
Encompass_Preferred
Fluorescein diacetate 5-?maleimide, AdipoGen Life Sciences. Chemical. CAS: 150322-01-3. Formula: C28H17NO9. MW: 511.4. Synthetic. Fluorescent marker used extensively in microscopy studies. Can be used in photoluminometric immunoassays for the formation of conjugates with immunoreactants by fluorescent energy transfer.
Catalog No. 50-101-6190