Learn More
Medchemexpress LLC Fluorescein Diacetate | 596-09-8 | 99.8% | 250 MG
SDP
Catalog No. 5000414258
Encompass_Preferred
Fluorescein diacetate is a cell-permeable esterase-substrate, an acetylated derivative of the green fluorescent dye fluorescein. It acts as a fluorogenic substrate for human Pi-class glutathione S-transferase (hGSTP1-1) and is utilized as a fluorescent probe for vital staining. Its fluorescence activation is dependent on hGSTP1-1 activity, making it useful for evaluating inhibitors of GSTP1-1 in vitro.
  • Cell-permeable esterase-substrate
  • Fluorogenic substrate for hGSTP1-1
  • Acetylated derivative of fluorescein
  • Fluorescent probe for vital staining
  • Used for evaluating inhibitors of GSTP1-1 in vitro
Catalog No. 50-004-14258 Supplier Medchemexpress LLC Supplier No. 250MGHYD0719R
Fluorescein diacetate is a cell-permeable esterase-substrate, an acetylated derivative of the green fluorescent dye fluorescein. It acts as a fluorogenic substrate for human Pi-class glutathione S-transferase (hGSTP1-1) and is utilized as a fluorescent probe for vital staining. Its fluorescence activation is dependent on hGSTP1-1 activity, making it useful for evaluating inhibitors of GSTP1-1 in vitro.
  • Cell-permeable esterase-substrate
  • Fluorogenic substrate for hGSTP1-1
  • Acetylated derivative of fluorescein
  • Fluorescent probe for vital staining
  • Used for evaluating inhibitors of GSTP1-1 in vitro