Learn More
Medchemexpress LLC HY-D0719 5g Medchemexpress, Fluorescein Diacetate CAS:596-09-8 Purity:>98%
SDP
Catalog No. 501933176
Encompass_Preferred
Medchemexpress, HY-D0719 5g Fluorescein Diacetate CAS:596-09-8 Fluorescein diacetate is a cell permeable esterase-substrate. Fluorescein diacetate can be used as a fluorogenic substrate for hGSTP1-1. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-193-3176 Supplier Medchemexpress LLC Supplier No. HYD07195G
Medchemexpress, HY-D0719 5g Fluorescein Diacetate CAS:596-09-8 Fluorescein diacetate is a cell permeable esterase-substrate. Fluorescein diacetate can be used as a fluorogenic substrate for hGSTP1-1. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.