Learn More
Sigma Aldrich Fine Chemicals Biosciences fluorescein diacetate used10G
Catalog No. 502899523
Encompass
fluorescein diacetate (fDA) is a non-fluorescent molecule which is hydrolysed to fluorescent fluorescein in live cells. Dead cells cannot accumulate or hydrolyse fDA.
Catalog No. 50-289-9523 Supplier Sigma Aldrich Fine Chemicals Biosciences Supplier No. F737810G
Add to Cart
Add to Cart
fluorescein diacetate (fDA) is a non-fluorescent molecule which is hydrolysed to fluorescent fluorescein in live cells. Dead cells cannot accumulate or hydrolyse fDA.