Learn More
AdipoGen Fluorescein dibutyrate
SDP
Catalog No. 502560906
Encompass_Preferred
Chemical. CAS 7298-65-9. Formula C28H24O7. MW 472.49. Synthetic. Fluorogenic substrate for esterase and lipase.
Catalog No. 50-256-0906 Supplier AdipoGen Supplier No. CDXF0057G025
Chemical. CAS 7298-65-9. Formula C28H24O7. MW 472.49. Synthetic. Fluorogenic substrate for esterase and lipase.