Learn More
AdipoGen Fluorescein dibutyrate (5 g)
SDP
Catalog No. 501490397
Encompass_Preferred
Fluorescein dibutyrate (5 g), AdipoGen Life Sciences. Chemical. CAS: 7298-65-9. Formula: C28H24O7. MW: 472.49. Synthetic. Fluorogenic substrate for esterase and lipase.
Catalog No. 50-149-0397 Supplier AdipoGen Supplier No. CDXF0057G005
Fluorescein dibutyrate (5 g), AdipoGen Life Sciences. Chemical. CAS: 7298-65-9. Formula: C28H24O7. MW: 472.49. Synthetic. Fluorogenic substrate for esterase and lipase.