Learn More
Abcam Fluorescein dibutyrate, Fluorogenic esterase and lipase substrate, 5G
Catalog No. 502705179
Encompass_Preferred
MW 472.5 Da, Purity >98%. Fluorogenic esterase and lipase substrate. Achieve your results faster with highly validated, pure and trusted compounds.
The product is subject to the following: Abcam Restricted Use Statement
Catalog No. 50-270-5179 Supplier Abcam Supplier No. AB1454535G
MW 472.5 Da, Purity >98%. Fluorogenic esterase and lipase substrate. Achieve your results faster with highly validated, pure and trusted compounds.
The product is subject to the following: Abcam Restricted Use Statement