Learn More
AdipoGen Fluorescein dicaproate
SDP
Supplier:  AdipoGen CDXF0089G001
Encompass_Preferred
Chemical. CAS 7364-90-1. Formula C32H32O7. MW 528.59. Fluorescein dicaproate is a non-fluorescent hydrophobic fluorescein derivative that can pass through the cell membrane whereupon intracellular esterases hydrolyze the dicaproate group producing the highly fluorescent product fluorescein. The fluorescein molecules accumulate in cells. Fluorescein dicaproate is a fluorogenic substrate for C6 esterase that generates green fluorescence upon esterase cleavage. Spectral Data lambdaEx 497nM, lambdaEm 516nm. Extinction coefficient 80000 cm-1 M-1.
Catalog No. 50-256-0881