Learn More
Cayman Chemical Fluorescein di-β-Galactopyranoside, Purity: ≥95%, CAS: 17817-20-8, 5 mg
SDP
Catalog No. 502933694
Encompass_Preferred
A fluorogenic substrate for β-galactosidase; upon enzymatic cleavage by β-galactosidase, fluorescein is released and its fluorescence can be used to quantify β-galactosidase activity; ex/em = 490/514 nm, respectively
Catalog No. 50-293-3694 Supplier Cayman Chemical Supplier No. 268605 MG
A fluorogenic substrate for β-galactosidase; upon enzymatic cleavage by β-galactosidase, fluorescein is released and its fluorescence can be used to quantify β-galactosidase activity; ex/em = 490/514 nm, respectively