Learn More
AdipoGen Fluorescein dilaurate
SDP
Catalog No. 502560896
Encompass_Preferred
Chemical. CAS 7308-90-9. Formula C44H56O7. Molecular Weight 696.91. Fluorescein dilaurate is a fluorogenic substrate for esterases/lipases and used in the pancreolauryl test for chronic pancreatitis. Spectral data lambdaex 490 nm lambdaem 514 nm in 0.1 M Tris pH 8.0 esterase.
Catalog No. 50-256-0896 Supplier AdipoGen Supplier No. CDXF0078G005
Chemical. CAS 7308-90-9. Formula C44H56O7. Molecular Weight 696.91. Fluorescein dilaurate is a fluorogenic substrate for esterases/lipases and used in the pancreolauryl test for chronic pancreatitis. Spectral data lambdaex 490 nm lambdaem 514 nm in 0.1 M Tris pH 8.0 esterase.