Learn More
AdipoGen Fluorescein dioctadecanoate
SDP
Catalog No. 502560874
Encompass_Preferred
Chemical. CAS 920269-51-8. Formula C56H80O7. MW 865.23. Fluorescein dioctadecanoate is a non-fluorescent hydrophobic fluorescein derivative that can pass through the cell membrane whereupon intracellular esterases hydrolyze the dioctadecanoate group producing the highly fluorescent product fluorescein. The fluorescein molecules accumulate in cells. Fluorescein dioctadecanoate is a fluorogenic substrate for C18 esterase that generates green fluorescence upon esterase cleavage. Spectral Data lambdaEx 497nM, lambdaEm 516nm. Extinction coefficient 80000 cm-1 M-1.
Catalog No. 50-256-0874 Supplier AdipoGen Supplier No. CDXF0094G001
Chemical. CAS 920269-51-8. Formula C56H80O7. MW 865.23. Fluorescein dioctadecanoate is a non-fluorescent hydrophobic fluorescein derivative that can pass through the cell membrane whereupon intracellular esterases hydrolyze the dioctadecanoate group producing the highly fluorescent product fluorescein. The fluorescein molecules accumulate in cells. Fluorescein dioctadecanoate is a fluorogenic substrate for C18 esterase that generates green fluorescence upon esterase cleavage. Spectral Data lambdaEx 497nM, lambdaEm 516nm. Extinction coefficient 80000 cm-1 M-1.