Learn More
AdipoGen Fluorescein dipalmitate
SDP
Catalog No. 502560877
Encompass_Preferred
Chemical. CAS 7262-38-6. Formula C52H72O7. MW 809.12. Fluorescein dipalmitate is a non-fluorescent hydrophobic fluorescein derivative that can pass through the cell membrane whereupon intracellular esterases hydrolyze the dipalmitate group producing the highly fluorescent product fluorescein. The fluorescein molecules accumulate in cells. Fluorescein dipalmitate is a fluorogenic substrate for C16 esterase that generates green fluorescence upon esterase cleavage. Spectral Data lambdaEx 497nM, lambdaEm 516nm. Extinction coefficient 80000 cm-1 M-1.
Catalog No. 50-256-0877 Supplier AdipoGen Supplier No. CDXF0093G001
Chemical. CAS 7262-38-6. Formula C52H72O7. MW 809.12. Fluorescein dipalmitate is a non-fluorescent hydrophobic fluorescein derivative that can pass through the cell membrane whereupon intracellular esterases hydrolyze the dipalmitate group producing the highly fluorescent product fluorescein. The fluorescein molecules accumulate in cells. Fluorescein dipalmitate is a fluorogenic substrate for C16 esterase that generates green fluorescence upon esterase cleavage. Spectral Data lambdaEx 497nM, lambdaEm 516nm. Extinction coefficient 80000 cm-1 M-1.